missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP10 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | BMP10 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16822115
|
Proteintech
22858-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16812115
|
Proteintech
22858-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
BMP10 is a member of the TGF-beta family of growth factors. Data suggest that the similar protein in mouse plays an important role in trabeculation of the embryonic heart. In human, this protein may signal through receptor serine/threonine kinases.Specifications
BMP10 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O95393 | |
27302 | |
BMP10 Fusion Protein Ag18746 | |
Primary | |
-20°C | |
BMP10 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BMP 10, BMP10, bone morphogenetic protein 10 | |
BMP10 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |