missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12492-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5′ untranslated region of this gene has been described and three variants are described, all encoding an identical protein.Specifications
BMP4 | |
Polyclonal | |
Unconjugated | |
BMP4 | |
BMP 2B, BMP 4, BMP2B, BMP2B1, BMP4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, ZYME | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12159, 25296, 652 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P12644, P21275, Q06826 | |
BMP4 | |
BMP4 Fusion Protein Ag3191 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |