missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ BMP9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA511931
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
BMP9 Polyclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in rodents suggest that this protein plays a role in the adult liver and in differentiation of cholinergic central nervous system neurons.
Specifikationer
| BMP9 | |
| Polyclonal | |
| Unconjugated | |
| GDF2 | |
| BMP; BMP9; BMP-9; Bone morphogenetic protein; Bone morphogenetic protein 9; Gdf2; GDF-2; growth differentiation factor 2; growth/differentiation factor 2; HHT5 | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 12165, 2658 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9UK05, Q9WV56 | |
| GDF2 | |
| KLH conjugated synthetic peptide between 8-37 amino acids from the N-terminal region of human BMP9 (GDF2). | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering