missing translation for 'onlineSavingsMsg'
Läs mer

BMS-1, MedChemExpress

Produktkod. 16470468
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
Förpackningsstorlek:
25mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16470468 5 mg -
16438008 10 mg -
16437786 100 mg -
16403937 50 mg 50mg
16403736 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16470468 Leverantör Medchem Express Leverantörsnummer HY19991

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress BMS-1 is an inhibitor of the PD-1/PD-L1 protein/protein interaction (IC50 between 6 and 100 nM).

BMS-1 is an inhibitor of the PD-1/PD-L1 protein/protein interaction (IC50 between 6 and 100 nM).

Specifikationer

Kemiskt namn eller material BMS-1
CAS 1675201-83-8
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C29H33NO5
Kvantitet 5 mg
Synonym PD-1/PD-L1 inhibitor 1
Löslighetsinformation Methanol : 25 mg/mL (52.57 mM; Need ultrasonic) ∣DMSO : 9 mg/mL (18.92 mM; Need ultrasonic and warming) ∣DMF : 5 mg/mL (10.51 mM; Need ultrasonic)
LEDER O=C([C@H]1N(CC2=C(OC)C=C(OCC3=C(C)C(C4=CC=CC=C4)=CC=C3)C=C2OC)CCCC1)O
Molekylvikt (g/mol) 475.58
Formel vikt 475.58
Procent renhet 98.58%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death
Mehr anzeigen Weniger anzeigen

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.