missing translation for 'onlineSavingsMsg'
Läs mer

BMS 299897, MedChemExpress

Produktkod. 16488128
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16488128 10 mg
16434047 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16488128 Leverantör Medchem Express Leverantörsnummer HY50883

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress BMS 299897 is a sulfonamide γ-secretase inhibitor with an IC50 of 7 nM for Aβ production inhibition in HEK293 cells stably overexpressing amyloid precursor protein (APP).

BMS 299897 is a sulfonamide γ-secretase inhibitor with an IC50 of 7 nM for Aβ production inhibition in HEK293 cells stably overexpressing amyloid precursor protein (APP).

Specifikationer

Kemiskt namn eller material BMS 299897
CAS 290315-45-6
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C24H21ClF3NO4S
Kvantitet 10 mg
Löslighetsinformation DMSO : ≥ 30 mg/mL (58.60 mM)
LEDER FC1=CC=C(F)C(N([C@@H](C2=C(CCCC(O)=O)C=C(F)C=C2)C)S(C3=CC=C(Cl)C=C3)(=O)=O)=C1
Molekylvikt (g/mol) 511.94
Formel vikt 511.94
Procent renhet 99.24%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.