Läs mer
BMS-345541, MedChemExpress
Produktkod. 16440377
Encompass_Preferred
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16440377 50 mg
16426737 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16440377 Leverantör Medchem Express Leverantörsnummer HY10519
MedChemExpress BMS-345541 is a selective inhibitor of the catalytic subunits of IKK (IKK-2 IC50=0.3 μM, IKK-1 IC50=4 μM). BMS-345541 binds at an allosteric site of IKK.
BMS-345541 is a selective inhibitor of the catalytic subunits of IKK (IKK-2 IC50=0.3 μM, IKK-1 IC50=4 μM). BMS-345541 binds at an allosteric site of IKK.
Specifikationer
Kemiskt namn eller material BMS-345541
Fysisk form Solid
CAS 445430-58-0
Färg White
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C14H17N5
För användning med (applikation) Cancer-Kinase/protease
Löslighetsinformation DMSO : 10 mg/mL (39.17 mM; Need ultrasonic)
LEDER CC1=CN=C2C(NCCN)=NC3=CC=C(C)C=C3N21
Molekylvikt (g/mol) 255.32
Formel vikt 255.32
Procent renhet 99.57%