missing translation for 'onlineSavingsMsg'
Läs mer

Tocris Bioscience™ BMS 466442
GREENER_CHOICE

Produktkod. 18796106
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18796106 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18796106 Leverantör Tocris Bioscience™ Leverantörsnummer 6878/10

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Potent and selective inhibitor of asc-1

BMS 466442 is a potent and selective asc-1 (alanine-serine-cysteine-1) transporter inhibitor (IC50 values are 20 and 37 nM in rat primary cortical cultures and HEK cells expressing asc-1 respectively). Displays >1000-fold selectivity for asc-1 over LAT-2 and ASCT-2. This compound is unsuitable for in vivo studies.
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material N-[[5-Methoxy-6-(phenylmethoxy)-1 H-indol-2-yl]carbonyl]-1-(phenylmethyl)-L-histidine methyl ester
CAS 1598424-76-0
Kvantitet 10 mg
Mål Other Ion Pump/Transporter Inhibitors
Molekylformel C31H30N4O5
Renhet 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.