missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMX Recombinant Human Protein
BMX Recombinant Human Protein
Brand: Invitrogen PV3371
Additional Details : Weight : 0.05250kg
Description
Bone marrow tyrosine kinase (BMX) is a non-receptor tyrosine kinase that belongs to the SRC-related TEC subfamily of tyrosine kinases. It is an important regulator of various cellular processes including apoptosis, cell survival, cellular differentiation, cell migration, and transformation.Specifications
NP_001712 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
83kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
BMX, ETK, PSCTK3, PSCTK2 | |
BMX was pre-diluted in enzyme dilution buffer and assayed in 12.5mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β- glycerophosphate, 2mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 and trace [32P]-γ-ATP for 10 minutes at 30°C | |
BMX Recombinant Human Protein | |
Human | |
Baculovirus | |
Cytoplasmic tyrosine kinase |
0.44mg/mL | |
660 | |
BMX Recombinant Human Protein | |
–80°C | |
1 Each | |
BMX Recombinant Human Protein | |
TEC | |
240nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1μg/mL | |
His Tag | |
Kinase and inhibitors | |
75% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.