missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ BNIP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586586
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Purity is > 95% by SDS-PAGE.
NIP3 is a member of the BCL2/adenovirus E1B 19 kd-interacting protein (BNIP) family. It interacts with the E1B 19 kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19 kDa-like sequences of BCL2, also an apoptotic protector. NIP3 contains a BH3 domain and a transmembrane domain, which have been associated with pro-apoptotic function. The dimeric mitochondrial protein is known to induce apoptosis, even in the presence of BCL2.
Specifikationer
| BNIP3 | |
| Polyclonal | |
| Unconjugated | |
| BNIP3 | |
| BCL2 interacting protein 3; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3; BCL2/adenovirus E1B 19 kDa-interacting protein 1, NIP3; BCL2/adenovirus E1B 19 kDa-interacting protein 3; BCL2/adenovirus E1B 19kDa interacting protein 3; BCL2/adenovirus E1B 19kDa-interacting protein 1, NIP3; BCL2/adenovirus E1B interacting protein 1, NIP3; BCL2/adenovirus E1B interacting protein 3; Bnip3; NIP3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12176, 664, 84480 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O55003, Q12983 | |
| BNIP3 | |
| Synthetic peptide, corresponding to amino acids 70-120 of Human BNIP-3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering