Learn More
Uvex BOA™ Black/Orange Rubber/PU 65312 Low Shoe - Width 11
Incorporates shock absorption, ergonomic design and low weight materials to ensure max performance, lightweight PU midsole with VIBRAM rubber outsole for high levels of cut, chemical and heat resistance, improved grip on slippery terrain
1985.00 SEK
Specifications
Color | Black, Orange |
---|---|
Material | Leather |
Sole Material | Dual-density polyurethane rubber (PU/RU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17201561
|
Uvex
6531235 |
35 |
1985.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17211561
|
Uvex
6531236 |
36 |
1985.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17221561
|
Uvex
6531237 |
37 |
1985.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17231561
|
Uvex
6531238 |
38 |
1985.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17241561
|
Uvex
6531239 |
39 |
1985.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17251561
|
Uvex
6531240 |
40 |
1985.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17261561
|
Uvex
6531241 |
41 |
1985.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17271561
|
Uvex
6531242 |
42 |
1985.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17281561
|
Uvex
6531243 |
43 |
1985.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17291561
|
Uvex
6531244 |
44 |
1985.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17201571
|
Uvex
6531245 |
45 |
1985.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17211571
|
Uvex
6531246 |
46 |
1985.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17221571
|
Uvex
6531247 |
47 |
1985.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17231571
|
Uvex
6531248 |
48 |
1985.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17241571
|
Uvex
6531249 |
49 |
1985.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17251571
|
Uvex
6531250 |
50 |
1985.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17261571
|
Uvex
6531251 |
51 |
1985.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17271571
|
Uvex
6531252 |
52 |
1985.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
Description
Uvex 2 VIBRAM Shoe with BOA™ Fit System 6531.2 S3 HI HRO SRC
- Sporty, multifunctional safety shoes
- Extremely light and flexible
- 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good side and ankle twist stability, no thermal conductivity and anti-magnetic properties
- Metal-free, penetration-resistance insole in accordance with latest standard, which does not impact shoe flexibility
- Made with new high-performance technology for exceptional wearer comfort
- Uvex Climazone technology for optimum comfort
- Breathable, water-resistant leather upper material
- Distance mesh lining provides optimum breathability
- Reflecting elements for more safety
- BOA™ Fit System Dial for custom comfort that is consistent all day
- Virtually seam-free construction to eliminate pressure points
- Removable comfort insole, antistatic, with moisture control system and additional shock absorption in heel and front of foot
- Soft, padded collar and tongue, dust tongue
- Injection-molded polyurethane scuffcap to protect upper leather on toe cap and significantly extend resistance time
- Meets ESD requirements: resistance to ground Kleiner 35 megohms
- Lightweight two-layer insole with excellent shock absorption characteristics and outstanding slip resistance
- Multilayer sole system comprising PU midsole and highly slip resistant rubber outsole from VIBRAM
- Optimized ankle protection to ensure safety when working on ladders
- Rugged, self-cleaning sole ridges ensure max grip or demanding applications on uneven or dirty floors (SRC labeling)
- Withstands brief exposure to heat approx. 300°C
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 6531.2
- Standard EN ISO 20345:2011 S3 HI HRO SRC
- Sizes: 35 to 52
Specifications
Black, Orange | |
Dual-density polyurethane rubber (PU/RU) | |
No | |
Unisex | |
Chemicals and heat | |
Yes | |
S3 |
Leather | |
Composite | |
EN ISO 20345:2011 | |
Standard width | |
Uvex 2 | |
No | |
SRC |