missing translation for 'onlineSavingsMsg'
Learn More

BOC-2-piperidylmethanol, 97%, ACROS Organics™

Brand:  Acros Organics 437520010

Additional Details : CAS Number : 157634-00-9 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 11976381

  • 416.00 SEK / 1g

Estimated Shipment: 11-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

157634-00-9
215.29
n-boc-piperidine-2-methanol, tert-butyl 2-hydroxymethyl piperidine-1-carboxylate, 1-boc-2-piperidinemethanol, n-boc-2-piperidinemethanol, 2-hydroxymethyl-1-n-boc-piperidine, 1-boc-2-hydroxymethyl-piperidine, boc-2-piperidylmethanol, 1-boc-2-hydroxymethylpiperidine, n-boc-2-hydroxymethyl piperidine, 2-hydroxymethyl-piperidine-1-carboxylic acid tert-butyl ester
tert-butyl 2-(hydroxymethyl)piperidine-1-carboxylate
C11H21NO3
PZTAGFCBNDBBFZ-UHFFFAOYSA-N
2763853
CC(C)(C)OC(=O)N1CCCCC1CO

Specifications

BOC-2-Piperidylmethanol
Glass Bottle
74.4°C to 75.7°C
157634-00-9
C11H21NO3
PZTAGFCBNDBBFZ-UHFFFAOYSA-N
tert-butyl 2-(hydroxymethyl)piperidine-1-carboxylate
2763853
Crystalline Powder
Authentic
White
1g
96% min. (GC)
n-boc-piperidine-2-methanol, tert-butyl 2-hydroxymethyl piperidine-1-carboxylate, 1-boc-2-piperidinemethanol, n-boc-2-piperidinemethanol, 2-hydroxymethyl-1-n-boc-piperidine, 1-boc-2-hydroxymethyl-piperidine, boc-2-piperidylmethanol, 1-boc-2-hydroxymethylpiperidine, n-boc-2-hydroxymethyl piperidine, 2-hydroxymethyl-piperidine-1-carboxylic acid tert-butyl ester
CC(C)(C)OC(=O)N1CCCCC1CO
215.29
215.29
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BOC-2-Piperidylmethanol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents
Certificates