Läs mer
Boc-Ala-Osu, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Boc-Ala-Osu |
| CAS | 3392-05-0 |
| Molekylformel | C12H18N2O6 |
| Kvantitet | 1 g |
| Synonym | (S)-2,5-Dioxopyrrolidin-1-yl 2-((tert-butoxycarbonyl)amino)propanoate, 2,5-Dioxopyrrolidin-1-yl (tert-butoxycarbonyl)-L-alaninate, N-tert-Butoxycarbonyl-L-alanine N-hydroxysuccinimide ester, N-tert-Butyloxycarbonyl-L-alanine N-hydroxysuccinimide ester, NSC 334372, tert-Butoxycarbonyl-L-alanine succinimido ester, tert-Butyloxycarbonyl-L-alanine N-hydroxysuccinimide ester, tert-Butyloxycarbonyl-L-alanine-N-hydroxysuccinimide |
| InChI formel | InChI=1S/C12H18N2O6/c1-7(13-11(18)19-12(2,3)4)10(17)20-14-8(15)5-6-9(14)16/h7H,5-6H2,1-4H3,(H,13,18)/t7-/m0/s1 |
| LEDER | C[C@H](NC(OC(C)(C)C)=O)C(ON1C(CCC1=O)=O)=O |
| IUPAC-namn | (S)-2,5-Dioxopyrrolidin-1-yl 2-((tert-butoxycarbonyl)amino)propanoate; 2,5-Dioxopyrrolidin-1-yl (tert-butoxycarbonyl)-L-alaninate; N-tert-Butoxycarbonyl-L-alanine N-hydroxysuccinimide ester; N-tert-Butyloxycarbonyl-L-alanine N-hydroxysuccinimide ester; NSC 334372; tert-Butoxycarbonyl-L-alanine succinimido ester; tert-Butyloxycarbonyl-L-alanine N-hydroxysuccinimide ester; tert-Butyloxycarbonyl-L-alanine-N-hydroxysuccinimide |
| Molekylvikt (g/mol) | 286.28 |
| Formel vikt | 286.12 |