Läs mer
Boc-D-4-Iodophenylalanine, TRC
Produktkod. 30008969
Change view
Kvantitet:
500 mg
1 g
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008969 2.5 g
30008207 500 mg
30003901 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008969 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6589482.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 176199-35-2
Molekylformel C14 H18 I N O4
Molekylvikt (g/mol) 391.202
IUPAC-namn (2R)-3-(4-iodophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
LEDER CC(C)(C)OC(=O)N[C@H](Cc1ccc(I)cc1)C(=O)O
Specifikationer
Kemiskt namn eller material Boc-D-4-Iodophenylalanine
InChI formel InChI=1S/C14H18INO4/c1-14(2,3)20-13(19)16-11(12(17)18)8-9-4-6-10(15)7-5-9/h4-7,11H,8H2,1-3H3,(H,16,19)(H,17,18)/t11-/m1/s1
Kvantitet 2.5 g
Formel vikt 391.028