missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOC-D-α-cyclohexylglycine, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Specifications
Assay Percent Range | 98% |
Packaging | Glass Bottle |
Formula Weight | 257.33 |
Percent Purity | 98% |
Quantity | 1 g |
Chemical Name or Material | BOC-D-α-Cyclohexylglycine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'