Läs mer
Boc-Gln-ONp, TRC
Produktkod. 30342837
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30342837 5 g
1 options
Produktkod. 30342837 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB3917955G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Boc-Gln-ONp
CAS 15387-45-8
Molekylformel C16H21N3O7
Kvantitet 5 g
Synonym N2-tert-Butoxycarbonyl-L-glutamine p-nitrophenyl ester, N2-(tert-Butyloxycarbonyl)-L-glutamine p-nitrophenyl ester, N-(tert-Butoxycarbonyl)-L-glutamine p-nitrophenyl ester, L-Boc-glutamine p-nitrophenyl ester
InChI formel InChI=1S/C16H21N3O7/c1-16(2,3)26-15(22)18-12(8-9-13(17)20)14(21)25-11-6-4-10(5-7-11)19(23)24/h4-7,12H,8-9H2,1-3H3,(H2,17,20)(H,18,22)/t12-/m0/s1
LEDER CC(C)(C)OC(=O)N[C@@H](CCC(=O)N)C(=O)Oc1ccc(cc1)[N+](=O)[O-]
IUPAC-namn (4-nitrophenyl) (2S)-5-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoate
Molekylvikt (g/mol) 367.35
Formel vikt 367.138