Läs mer
Boc-L-cysteine, TRC
Produktkod. 30179586
Change view
Kvantitet:
250 mg
1 g
2 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179586 2 g
30179114 250 mg
30179318 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179586 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6193702G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 20887-95-0
Molekylformel C8H15NO4S
Molekylvikt (g/mol) 221.27
Synonym N-[(1,1-Dimethylethoxy)carbonyl]-L-cysteine, N-Carboxy-L-cysteine N-tert-Butyl Ester, (R)-2-tert-Butoxycarbonylamino-3-mercaptopropionic Acid, N-(tert-Butoxycarbonyl)cysteine, N-(tert-Butyloxycarbonyl)-L-cysteine, N-BOC-L-Cysteine, N-tert-Butoxycarbonyl-L-cysteine
IUPAC-namn (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-sulfanylpropanoic acid
LEDER CC(C)(C)OC(=O)N[C@@H](CS)C(=O)O
Specifikationer
Kemiskt namn eller material Boc-L-cysteine
InChI formel InChI=1S/C8H15NO4S/c1-8(2,3)13-7(12)9-5(4-14)6(10)11/h5,14H,4H2,1-3H3,(H,9,12)(H,10,11)/t5-/m0/s1
Kvantitet 2 g
Formel vikt 221.0722