Läs mer
Boc-Lys(Z)-OtBu N-Phenylmethoxycarbonyl, TRC
Produktkod. 30329434
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30329434 500 mg
1 options
Produktkod. 30329434 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB657075500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Boc-Lys(Z)-OtBu N-Phenylmethoxycarbonyl
CAS 135727-85-4
Molekylformel C23H36N2O6
Kvantitet 500 mg
Synonym (S)-Tert-butyl 6-(((Benzyloxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)hexanoate, N2-[(1,1-Dimethylethoxy)carbonyl]-N6-[(phenylmethoxy)carbonyl]-L-lysine 1,1-Dimethylethyl Ester
InChI formel InChI=1S/C23H36N2O6/c1-22(2,3)30-19(26)18(25-21(28)31-23(4,5)6)14-10-11-15-24-20(27)29-16-17-12-8-7-9-13-17/h7-9,12-13,18H,10-11,14-16H2,1-6H3,(H,24,27)(H,25,28)
LEDER O=C(OCC=1C=CC=CC1)NCCCCC(NC(=O)OC(C)(C)C)C(=O)OC(C)(C)C
IUPAC-namn tert-butyl N6-((benzyloxy)carbonyl)-N2-(tert-butoxycarbonyl)lysinate
Molekylvikt (g/mol) 436.26
Formel vikt 436.54