Läs mer
MedChemExpress Boc-NH-PEG2
Produktkod. 16479316
Encompass_Preferred
Change view
100 mg
Produktkod.
16479316 100 mg
1 options
Produktkod. 16479316 Leverantör MedChemExpress Leverantörsnummer HYW004896
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
MedChemExpress Boc-NH-PEG2 (PROTAC Linker 11) is a PEG-based PROTAC linker can be used in the synthesis of PROTACs.
Specifikationer
Molekylvikt (g/mol) 205.25 g/mol
CAS 139115-91-6
Innehåll och lagring Pure form -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Frakt skick Room temperature
Kvantitet 100 mg
Synonym PROTAC Linker 11
Mål NO Synthase ∣ MMP ∣ Endogenous Metabolite