Learn More
BOC-trans-4-hydroxy-L-proline, 98%, ACROS Organics™
Catalog Number p-4228410
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
13726-69-7
C10H17NO5
231.25
MFCD00053370
BENKAPCDIOILGV-UHFFFAOYNA-N
boc-hyp-oh, boc-l-hydroxyproline, 2s,4r-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, boc-trans-4-hydroxy-l-proline, n-boc-trans-4-hydroxy-l-proline, boc-l-hyp-oh, boc-hydroxyproline, unii-rc99p5b8ig, trans-boc-hyp-oh, 4r-1-tert-butoxycarbonyl-4-hydroxy-l-proline
88804
1-[(tert-butoxy)carbonyl]-4-hydroxypyrrolidine-2-carboxylic acid
CC(C)(C)OC(=O)N1CC(O)CC1C(O)=O
13726-69-7
C10H17NO5
231.25
MFCD00053370
BENKAPCDIOILGV-UHFFFAOYNA-N
boc-hyp-oh, boc-l-hydroxyproline, 2s,4r-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, boc-trans-4-hydroxy-l-proline, n-boc-trans-4-hydroxy-l-proline, boc-l-hyp-oh, boc-hydroxyproline, unii-rc99p5b8ig, trans-boc-hyp-oh, 4r-1-tert-butoxycarbonyl-4-hydroxy-l-proline
88804
1-[(tert-butoxy)carbonyl]-4-hydroxypyrrolidine-2-carboxylic acid
CC(C)(C)OC(=O)N1CC(O)CC1C(O)=O
Melting Point 122.0°C to 123.4°C
Color White
Elemental Analysis 50.6 to 53.2% C, 5.9 to 6.2% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Specific Rotation -51.5° to -55.5° (20°C, 589nm) (c=1 in DMF)
Formula Weight 231.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material BOC-trans-4-Hydroxy-L-proline