Läs mer
Boceprevir Metabolite M4-d9 Methyl Ester, TRC
Produktkod. 30338863
Change view
Click to view available options
Kvantitet:
0.5 mg
Unit Size:
0.5mg
Product Code. Kvantitet unitSize
30338863 0.5 mg 0.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30338863 Supplier Toronto Research Chemicals Supplier No. TRCB6745170.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Boceprevir metabolite M4 tert-butyl-d9 methyl ester
CAS 1158083-48-7
Molekylformel C20 2H9 H26 N3 O4
Kvantitet 0.5 mg
Synonym (1R,2S,5S)-3-[(2S)-2-[[[[1,1-Di(methyl-d3)ethyl-2,2,2-d3]amino]carbonyl]amino]-3,3-di(methyl-d3)-1-oxobutyl-4,4,4-d3]-6,6-dimethyl-3-azabicyclo[3.1.0]hexane-2-carboxylic Acid Methyl Ester
InChI formel InChI=1S/C20H35N3O4/c1-18(2,3)14(21-17(26)22-19(4,5)6)15(24)23-10-11-12(20(11,7)8)13(23)16(25)27-9/h11-14H,10H2,1-9H3,(H2,21,22,26)/t11-,12-,13-,14+/m0/s1/i4D3,5D3,6D3
LEDER [2H]C([2H])([2H])C(NC(=O)N[C@H](C(=O)N1C[C@H]2[C@@H]([C@H]1C(=O)OC)C2(C)C)C(C)(C)C)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC-namn methyl (1R,2S,5S)-3-[(2S)-2-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]carbamoylamino]-3,3-dimethylbutanoyl]-6,6-dimethyl-3-azabicyclo[3.1.0]hexane-2-carboxylate
Molekylvikt (g/mol) 390.57
Formel vikt 390.3192