Learn More
Velp Scientifica™ BOD Bottle
Product Code. 11716913 Shop All Velp Products
Change view
Click to view available options
Volume (Metric):
500 mL
Unit Size:
Each
Product Code. Volume (Metric) Unit Size
11716913 500 mL Each
1 options
Product Code. 11716913 Supplier Velp Scientifica™ Supplier No. 10001107
  • ONLINE EXCLUSIVE PRICE
    133.00 SEK
    129.08 SEK / Paket med 1
    Save 3.92 SEK
    2% Av
I lager
Velp Scientifica™ BOD Bottle is a spare part for BOD and BOD EVO sensors.
  • Constructed of glas
  • Dark in color
Specifications
Volume (Metric) 500 mL
For Use With (Equipment) BOD and BOD EVO sensor