Läs mer
WTW™ BOD OxiTop™ Thermostat Box
Vikt (metrisk):
30 kg
Förpackningsstorlek:
Varje
Beskrivning
- Thermoboxes to incubate BSB samples (dilution samples or respirometric samples) with constant temperature for simple requirements
- Space-saving and compact
- Convection cooling for even temperature distribution
- Thermostat box with convection cooling for BSB OxiTop™ measuring systems with max. 12 measuring points or 20 Karlsruhe bottles for simple requirements
- Standard factory setting at 20 ° C. Low-maintenance with automatic dew water evaporation
Specifikationer
Specifikationer
| Typ | BOD OxiTop™ Thermostat Box |
| Temperaturnoggrannhet | 20°C |
| Temperatur (metrisk) Drift | 10°C to 32°C |
| Inkluderar | Temperature-controlled forced ventilation, hinged non-corrosive clear-view cover |
| Garanti | 18 Months |
| Bredd (metrisk) | 425 mm |
| Höjd (metrisk) | 375 mm |
| Djup (metriskt) | 600 mm |
| Vikt (metrisk) | 30 kg |