Läs mer
Boldine, TRC
Produktkod. 30136302
Change view
Kvantitet:
1 g
2.5 g
5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30136302 2.5 g 2.5g
30135311 5 g -
30008981 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136302 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6751702.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 476-70-0
Molekylformel C19 H21 N O4
Molekylvikt (g/mol) 327.37
Synonym 4H-Dibenzo[de,g]quinoline-2,9-diol, 5,6,6a,7-tetrahydro-1,10-dimethoxy-6-methyl-, (6aS)-, 4H-Dibenzo[de,g]quinoline-2,9-diol, 5,6,6a,7-tetrahydro-1,10-dimethoxy-6-methyl-, (S)-, Boldine (6CI,7CI), (+)-(S)-Boldine, (+)-2,9-Dihydroxy-1,10-dimethoxyaporphine, (+)-Boldine, (S)-(+)-Boldine, (S)-Boldine, Boldin, Boldina, NSC 65689, d-Boldine, 6aalpha-Aporphine-2,9-diol, 1,10-dimethoxy- (8CI), 1,10-Dimethoxy-6aalpha-aporphine-2,9-diol
IUPAC-namn (6aS)-1,10-dimethoxy-6-methyl-5,6,6a,7-tetrahydro-4H-dibenzo[de,g]quinoline-2,9-diol
LEDER COc1cc2c(C[C@@H]3N(C)CCc4cc(O)c(OC)c2c34)cc1O
Specifikationer
Kemiskt namn eller material Boldine
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 g
InChI formel InChI=1S/C19H21NO4/c1-20-5-4-10-7-15(22)19(24-3)18-12-9-16(23-2)14(21)8-11(12)6-13(20)17(10)18/h7-9,13,21-22H,4-6H2,1-3H3/t13-/m0/s1
Formel vikt 327.1471
Procent renhet >95%