Learn More
Borane-2-methylpyridine complex, 95%, Thermo Scientific Chemicals
Description
Used as reactant for N-Benzyl-protection of amino acid derivatives by reductive alkylation, reductive alkoxyamination, as reducing agent for the labeling of oligosaccharides by reductive amination, synthesis of alkoxyamine derivatives, via reduction of oxime ethers, reductive amination reactions of C1-C10 aldehyde 2,4-dinitrophenylhydrazones, synthesis of various trifluoromethylated amino compounds and an alternative reagent for reductive aminations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used as reactant for N-Benzyl-protection of amino acid derivatives by reductive alkylation, reductive alkoxyamination, as reducing agent for the labeling of oligosaccharides by reductive amination, synthesis of alkoxyamine derivatives, via reduction of oxime ethers, reductive amination reactions of C1-C10 aldehyde 2,4-dinitrophenylhydrazones, synthesis of various trifluoromethylated amino compounds and an alternative reagent for reductive aminations.
Solubility
Slightly sol. in water, cyclohexane, very sol. in methanol, THF, diglyme, and benzene
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Melting Point | 44°C to 50°C |
Color | White |
Flash Point | 100°C (212°F) |
Quantity | 25 g |
UN Number | UN2813 |
Sensitivity | Moisture sensitive |
Solubility Information | Slightly sol; in water,cyclohexane,very sol; in methanol,THF,diglyme,and benzene |
Formula Weight | 106.96 |
Percent Purity | 95% |
Physical Form | Solid |
Show More |
RUO – Research Use Only