Läs mer
(+)-Bornyl Acetate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (+)-Bornyl Acetate |
| CAS | 20347-65-3 |
| Molekylformel | C12 H20 O2 |
| Kvantitet | 5 g |
| Synonym | Bicyclo[2.2.1]heptan-2-ol, 1,7,7-trimethyl-, 2-acetate, (1R,2S,4R)-, 2-Bornanol, acetate, (1R,2S,4R)-(+)- (8CI), Bicyclo[2.2.1]heptan-2-ol, 1,7,7-trimethyl-, acetate, (1R,2S,4R)- (9CI), Bicyclo[2.2.1]heptan-2-ol, 1,7,7-trimethyl-, acetate, (1R-endo)-, Borneol, d-, acetate (6CI), (+)-Borneol acetate, (+)-Bornyl, acetate, d-Bornyl acetate |
| InChI formel | InChI=1S/C12H20O2/c1-8(13)14-10-7-9-5-6-12(10,4)11(9,2)3/h9-10H,5-7H2,1-4H3/t9-,10+,12+/m1/s1 |
| LEDER | CC(=O)O[C@H]1C[C@H]2CC[C@]1(C)C2(C)C |
| IUPAC-namn | [(1R,3S,4R)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] acetate |
| Molekylvikt (g/mol) | 196.29 |
| Formel vikt | 196.1463 |