Läs mer
Boron Subphthalocyanine Chloride, TRC
Produktkod. 30344220
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30344220 500 mg
1 options
Produktkod. 30344220 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB675418500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Boron Subphthalocyanine Chloride
CAS 36530-06-0
Molekylformel C24H12BClN6
Kvantitet 500 mg
Synonym (T-4)-Chloro[7,12:14,19-diimino-21,5-nitrilo-5H-tribenzo[c,h,m][1,6,11]triazacyclopentadecinato(2-)-N22,N23,N24]boron7,12:14,19-Diimino-21,5-nitrilo-5H-tribenzo[c,h,m][1,6,11]triazacyclopentadecine, Boron Complex, 7,12:14,19-Diimino-21,5-nitrilo-5H-tribenzo[c,h,m][1,6,11]triazacyclopentadecine Boron Deriv., Boron Subphthalocyanine Chloride, Boron(III) Subphthalocyanine Chloride, Chloro(subphthalocyaninato)boron, Chloroboronsubphthalocyanine
InChI formel InChI=1S/C24H12BClN6/c26-25-30-19-13-7-1-2-8-14(13)20(30)28-22-17-11-5-6-12-18(17)24(32(22)25)29-23-16-10-4-3-9-15(16)21(27-19)31(23)25/h1-12H
LEDER Cl[B]12N3C4=NC5=[N]1C(=Nc6c7ccccc7c(N=C3c8ccccc48)n26)c9ccccc59
IUPAC-namn 12-chloro-2,11,13,22,30,31-hexaza-12-boraoctacyclo[21.6.1.110,14.03,11.04,9.013,21.015,20.024,29]hentriaconta-1,3,5,7,9,14(31),15,17,19,21,23(30),24,26,28-tetradecaene
Molekylvikt (g/mol) 430.66
Formel vikt 430.0905