Learn More
Robu™ Borosilicate Glass Buchner Filter Funnels
Brand: Robu™ 21752
Additional Details : Weight : 0.10000kg
Specifications
Buchner filter funnel | |
75 mL | |
Borosilicate Glass | |
Highly resistant to most chemicals such as solvents, acids and alkalis | |
2 |
52mm (OD); 48mm (ID) | |
10mm | |
50 mm | |
52mm |