Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Conical Filter Tube
Made of DURAN™ borosilicate glass. Lenz Laborglasinstrumente™ Borosilicate Glass Conical Filter Tubes are used for filtration of very small volumes and is highly resistant to most chemicals such as solvents, acids, and alkalis.
Brand: Lenz Laborglasinstrumente™ 07220004
Additional Details : Weight : 0.10000kg
Description
- Filter tubes
- Made of borosilicate glass
Specifications
Filter Tube | |
Borosilicate Glass | |
4 |
30 mL | |
20 mm |