Läs mer
Fisherbrand™ Borosilicate Glass Conical Shape Narrow Neck Erlenmeyer Flasks
Produktkod. 12917620 Handla allt Fisherbrand Traceable Produkter
Change view
Capacity: 250mL
Produktkod.
12917620 Capacity: 250mL
1 options
Produktkod. 12917620
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Erlenmeyer, graduated.
Specifikationer
Kapacitet (metrisk) 250 mL
Material Borosilicate Glass
Form Conical
Mun stil Narrow