Learn More
Cole-Parmer™ Borosilicate Glass Erlenmeyer Flasks
Ideal for culturing, mixing, and storing media
Brand: Cole-Parmer™ 34554-04
Additional Details : Weight : 0.80000kg
Description
- Impact-resistant caps have cemented-in, inert, rubber liners that are resistant to steam sterilization
- Graduated scale
- Marking area
Specifications
Phenolic with white rubber liner | |
Borosilicate Glass | |
50 mL | |
Screw cap | |
38-430M thread |
500 mL | |
100 to 500 mL | |
186 mm | |
Screw |