Learn More
Cole-Parmer™ Borosilicate Glass Erlenmeyer Flasks
Ideal for culturing, mixing, and storing media
Brand:  Cole-Parmer™ 34554-03
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 11800080
Quantity Price
1 2006.00 SEK / Pack of 6
6 1785.94 SEK / Pack of 6
Estimated Shipment
16-06-2021
Add to basket
Description
Description
  • Impact-resistant caps have cemented-in, inert, rubber liners that are resistant to steam sterilization
  • Graduated scale
  • Marking area
Specifications
Specifications
Phenolic with white rubber liner
Borosilicate Glass
25 mL
Screw cap
38-430M thread
250 mL
50 to 200 mL
135 mm
Screw