Learn More
Fisherbrand™ Borosilicate Mini Homogenizers
For use during the initial stages of sample preparation to break down and disrupt a range of materials such as animal or plant tissues, foodstuffs and soil. Fisherbrand™ Borosilicate Mini Homogenizers are tissue grinders manufactured from durable borosilicate glass.
Brand: Fisherbrand™
Additional Details : Weight : 0.03008kg
Description
- Ideal for micro tissue work
- Bodies are clear precision glass
- Pestles have ground glass working length
Specifications
Clear | |
Borosilicate glass | |
55mm |
1.0mL | |
5.0mm diameter |