Learn More
Quickfit™ Borosilicate Glass Markham Still Body
Brand:  Quickfit MC46/SC
4607.50 SEK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 1.78000kg
Product Code. 12455306
  • 4850.00 SEK / Each
Estimated Shipment: 23-08-2024
to see stock.
Specifications
Markham Still Body
25mL
S19
Borosilicate Glass
25mL