Learn More
Borosilicate Glass Petri Dishes
Constructed of borosilicate glass. Scientific Glass Labs™ Borosilicate Glass Petri Dishes are autoclavable and include lid.
Brand: Fisherbrand
Additional Details : Weight : 0.50000kg
Description
- Constructed of borosilicate glass.
- Includes lid
- Autoclavable
Specifications
Closure Included | |
Borosilicate Glass | |
Petri Dish | |
150 mm |
Borosilicate Glass | |
Round | |
25 mm |