Learn More
Robu™ Borosilicate Glass Standard Filter Crucibles
Brand:  Robu™ 20305
Additional Details : Weight : 0.33910kg
 View more versions of this product
Product Code. 16873006
  • 3335.00 SEK / Pack of 10
Estimated Shipment: 24-06-2024
to see stock.
Specifications
36mm
Analytical filtration in laboratories
36mm
Standard Filter Crucible
Borosilicate Glass
30 mL
56mm
5