Learn More
Quickfit™ Borosilicate Glass Stopcock Adapter with Sockets
Control flow rate. QuickFit™ borosilicate glass stopcock adater with cone comes with a GP Rotoflo™ stopcock.
1130.00 SEK - 5640.00 SEK
Specifications
Product Type | Stopcock Adapter |
---|---|
Material | Borosilicate Glass |
Includes | Sockets |
Product Code | Brand | Port Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Port Size | Price | Quantity & Availability | |||||
12415316
|
Quickfit
MF14/2/SC |
19/26 |
1130.00 SEK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
15567930
|
Quickfit
MF14/3/SC |
24/29 |
5640.00 SEK
Pack of 5 |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
15557930
|
Quickfit
MF14/1/SC |
14/23 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- With GP Rotaflo™ PTFE stopcock providing fine flow control and positive closure
- Detachable plastic screwthread connectors allow rubber hose etc. , to be fitted easily and safely
Specifications
Stopcock Adapter | |
Borosilicate Glass | |
Sockets |