Learn More
Karl Hecht™ Volumetric Flask with PP Stopper
Simply and accurately measure required sample volumes. Karl Hecht™ Volumetric Flask with PP Stopper is thick-walled, clearly marked and labeled, and has a solid finish.
4817.70 SEK
Product Code | Brand | Capacity (Metric) | Closure Size | Neck Style | Tolerance | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Neck Style | Tolerance | Price | Quantity & Availability | ||
10650012
|
Karl Hecht™
42094092 |
3000 mL | NS 29 | Standard | 1.2 mL |
4817.70 SEK
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
||
Description
- Duran™ borosilicate glass 3.3.
- Standard type.
- Fused blue graduation.
- Standard ground and interchangeable NS stopper of polyethylene.
- Thick-walled, solid finish.
- Clearly legible figures and marks.
Specifications
Yes |