Läs mer
Bortezomib, TRC
Produktkod. 30010791
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010791 100 mg
30006624 5 mg
30006475 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010791 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB675700100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 179324-69-7
Molekylformel C19 H25 B N4 O4
Molekylvikt (g/mol) 384.24
Synonym Boronic acid, B-[(1R)-3-methyl-1-[[(2S)-1-oxo-3-phenyl-2-[(2-pyrazinylcarbonyl)amino]propyl]amino]butyl]-, Boronic acid, [(1R)-3-methyl-1-[[(2S)-1-oxo-3-phenyl-2-[(pyrazinylcarbonyl)amino]propyl]amino]butyl]- (9CI), Boronic acid, [3-methyl-1-[[1-oxo-3-phenyl-2-[(pyrazinylcarbonyl)amino]propyl]amino]butyl]-, [S-(R*,S*)]-, Bortezomib, DPBA, LDP 341, MG 341, MLN 341, NSC 681239, PS 314, PS 341, PS 341 (pharmaceutical), Radiciol, Velcade
IUPAC-namn [(1R)-3-methyl-1-[[(2S)-3-phenyl-2-(pyrazine-2-carbonylamino)propanoyl]amino]butyl]boronic acid
LEDER CC(C)C[C@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)c2cnccn2)B(O)O
Specifikationer
Kemiskt namn eller material Bortezomib
Kvantitet 100 mg
InChI formel InChI=1S/C19H25BN4O4/c1-13(2)10-17(20(27)28)24-18(25)15(11-14-6-4-3-5-7-14)23-19(26)16-12-21-8-9-22-16/h3-9,12-13,15,17,27-28H,10-11H2,1-2H3,(H,23,26)(H,24,25)/t15-,17-/m0/s1
Formel vikt 384.1969