Learn More
Scientific Industries SI™ Bottom Closure with Feet Kit
Fits onto Vortex-Genie 1, Vortex-Genie Pulse, Disruptor Genie, and Vortex-Genie 2 Family of mixers. Scientific Industries SI™ Bottom Closure with Feet Kit includes screws and installs easily.
Brand:  Scientific Industries SI™ 0K-0236-408
Additional Details : Weight : 0.72000kg
Product Code. 15537315
  • 682.00 SEK / Each
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
Description
Description
  • Installs easily with nut driver or pliers
  • Includes four screws
Specifications
Specifications
Molded plastic
152.4 mm
Vortex-Genie
4 Screws