Learn More
SCAT Box for Politainer (Foldable Waste Container)
Catalog Number p-7236269
Click to view available options
Capacity (Metric):
5 L
10 L
20 L
Outer carton for the SCAT Politainer - also known as folding canisters.
SCAT Politainers are an ideal way of not having to dispense with safe storage even in limited spaces
Capacity (Metric) 5 L
Dimensions (L x W x H) 190 x 189 x 186 mm
For Use With (Application) Waste Disposal System
Color White (Translucent)
For Use With (Equipment) Politainer