Learn More
Trajan Scientific and Medical™ BP5MS GC Capillary Columns
Optimized silphenylene column for general purpose MS analysis.
Brand: Trajan Scientific and Medical™ 054320
Additional Details : Weight : 0.01000kg
Description
- Good for 5% GCMS analysis
- Optimized silphenylene content for general purpose MS analyses
Specifications
BP5MS GC Capillary Column | |
GC/MS | |
0.25 μm | |
5% Phenyl Polysilphenylene-siloxane |
0.32 mm | |
-40°C to +330/350°C | |
30 m | |
BP5MS |