Learn More
Trajan Scientific and Medical™ BPX-BIOD GC Capillary Columns
Provides optimal separation. Trajan Scientific and Medical BPX-BIOD GC Capillary Columns are optimized for specific ASTM and EN biodiesel methods.
Brand:  Trajan Scientific and Medical™ 0544332
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15268673
  • 5660.00 SEK / Each
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
  • Optimized for specified ASTM and EN biodiesel methods
Specifications
Specifications
Columns for EN14110
0.32mm
30m
BPX-BIOD GC Capillary Columns
0.25μm
BPX-BIOD Wax
Videos