Learn More
Trajan Scientific and Medical BPX5 Nonpolar GC Capillary Column
Product Code. 16662612
Change view
Click to view available options
Length (Metric):
6 m
10 m
12 m
15 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.1 mm
0.15 mm
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.15 μm
0.25 μm
0.5 μm
1 μm
Unit Size:
Each
30 product options available for selection
Product selection table with 30 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Inner Film Thickness Unit Size
16662612 60 m 0.32 mm 1 μm Each
15218653 6 m 0.32 mm 1 μm Each
10567342 12 m 0.22 mm 0.25 μm Each
10442341 25 m 0.32 mm 0.25 μm Each
10519452 12 m 0.53 mm 0.25 μm Each
12684945 60 m 0.25 mm 0.25 μm Each
16642612 30 m 0.53 mm 1 μm Each
16622612 25 m 0.53 mm 1 μm Each
10320941 50 m 0.22 mm 0.25 μm Each
10269190 25 m 0.53 mm 0.25 μm Each
15124079 30 m 0.53 mm 0.5 μm Each
10430081 25 m 0.32 mm 0.5 μm Each
10268810 25 m 0.22 mm 0.25 μm Each
10451031 25 m 0.15 mm 0.25 μm Each
10791412 30 m 0.22 mm 0.25 μm Each
10208670 60 m 0.32 mm 0.25 μm Each
10567532 12 m 0.32 mm 0.25 μm Each
10025450 12 m 0.53 mm 1 μm Each
10300711 12 m 0.32 mm 1 μm Each
15288643 15 m 0.25 mm 1 μm Each
10616021 30 m 0.32 mm 0.25 μm Each
10177200 15 m 0.32 mm 0.25 μm Each
12905561 15 m 0.25 mm 0.25 μm Each
10762562 50 m 0.32 mm 0.5 μm Each
15298643 30 m 0.25 mm 1 μm Each
10661813 10 m 0.1 mm 0.1 μm Each
10055400 50 m 0.32 mm 1 μm Each
15248643 30 m 0.15 mm 0.15 μm Each
15238643 30 m 0.25 mm 0.1 μm Each
15228663 15 m 0.25 mm 0.1 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 30 options available.
30 options
Product Code. 16662612 Supplier Trajan Scientific and Medical Supplier No. 054154
Beräknad leverans: 16-12-2026
Capillary column provides a great low-polarity option for GC analyses. Recommended for ultra trace analyses, pesticides, herbicides, hydrocarbons, solvents, phenols, and amines.
  • High temperature.
  • General purpose GC column (suitable for over 80% routine GC analyses).
  • Very low bleed – ideal for trace analysis.
  • Non-polar.
  • Extremely inert.
  • Ideal for GCMS.
Specifications
For Use With (Application) Amines Analysis, Pesticides and Herbicides Analysis, Hydrocarbons, Solvents Analysis, Phenols, Amines, GC/MS
Film Thickness 1 μm
Length (Metric) 60 m
Material Fused Silica
Packing Material BPX5
Polarity Non polar
Coating Fused Silica
USP Type G27, G36, G41
Stationary Phase 5% Phenyl Polysilphenylene Siloxane
Type GC Capillary Column
Diameter (Metric) Inner 0.32 mm
Functionality/Interaction Type Pi Bonding with the Aromatic Phenyl Groups
Max. Temperature (Metric) 370°C
Temperature Range (Metric) -40°C to +360/370°C