Learn More
Brachyury Monoclonal Antibody (X1AO2), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1360.00 SEK - 3195.00 SEK
Specifications
Antigen | Brachyury |
---|---|
Clone | X1AO2 |
Concentration | 0.5 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15547666
|
Affymetrix eBioscience
14-9770-80 |
25 μg |
1360.00 SEK
25µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15557666
|
Affymetrix eBioscience
14-9770-82 |
100 μg |
3195.00 SEK
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The monoclonal antibody X1AO2 recognizes human Brachyury, a 47 kDa protein encoded by the T gene and functioning as a nuclear transcription factor. This transcription factor binds to specific DNA regions near the palindromic sequence TCACACCT (T-site) through a region in its N-terminus, called the T-box. Brachyury functions during embryogenesis to regulate midline development by establishing the anterior/posterior axis through the regulation of genes involved in mesoderm formation and differentiation. Brachyury is the primary mesoderm marker used to distinguish differentiation of embryonic stem cells into a mesenchymal stem cell lineage. Variations in the T gene in humans are associated with neural tube defects and chordomas. Expression of brachyury is also increased in advanced stage lung tumors and tumor cell lines. Applications Reported: This X1AO2 antibody has been reported for use in western blotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, immunohistochemical staining, and immunocytochemistry. Applications Tested: This X1AO2 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded human tissue using low pH antigen retrieval and can be used at less than or equal to 5 μg/mL. This X1AO2 antibody has also been tested by immunocytochemistry of fixed and permeabilized human cells and can be used at less than or equal to 5 μg/mL.
The protein encoded by this gene is an embryonic nuclear transcription factor that binds to a specific DNA element, the palindromic T-site. It binds through a region in its N-terminus, called the T-box, and effects transcription of genes required for mesoderm formation and differentiation. The protein is localized to notochord-derived cells.Specifications
Brachyury | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Bra; brachyury; brachyury homolog; brachyury protein; brachyury-like 2; brachyury-like 3; cou; D17Mit170; Low; low ratio; Lr; me75; Protein T; RP11-459F1.1; SAVA; T; T brachyury homolog; T brachyury transcription factor; T, brachyury homolog; T1; T-box transcription factor T; TBXT; TFT; Tl2; Tl3 | |
TBXT | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
X1AO2 | |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
O15178 | |
6862 | |
E.coli-derived fragment of human Brachyury corresponding to amino acids 237-348 | |
Primary | |
4° C | |
TBXT |
For Research Use Only.