Learn More
Brady™ Accessories for BMP™51 and BMP™53 Label Printers
Accessories for BMP™51 and BMP™53 label printers.
575.00 SEK - 2285.00 SEK
Specifications
For Use With (Equipment) | BMP51 Label Printer, BMP53 Label Printer |
---|---|
Sterility | Non-sterile |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
11902742
|
Brady™
143109 |
Hard Case |
2285.00 SEK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
12773989
|
Brady™
143115 |
BMP50 Series Spare Battery Tray |
575.00 SEK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
11992732
|
Brady™
143108 |
Magnet Accessory |
1430.00 SEK
Each |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
Description
- BMP™51 and BMP™53 Printer Accessories
- Printer Cases designed for use with BMP™51 and BMP™53 Label Printers from Brady™ protects your printer against impacts and dust while allowing for easy mobility
- Printer Magnet Attachment allows printer to affix to panel door or metal surface for more hands-free operation or storage
- Attaches to back of printer with screws to ensure stability
- Printer Battery Tray fits 8 AA batteries
- User may pre-load batteries into the spare tray for quick changeover in the field
- The battery tray snaps in place into the battery compartment
Specifications
BMP51 Label Printer, BMP53 Label Printer | |
Non-sterile |