Läs mer
Brady™ BradyJet J2000 Colour Label Printer - EU with Brady Workstation LAB Suite
Beskrivning
The perfect combo. Get the impact of colour—that’s wildly durable—to create a safer, more efficient workplace. Rely on the BradyJet J2000 Colour Label Printer when durable colour matters most.
- Works great in a multitude of industries, from lab to safety and more
- Compact design means it can snuggle in to tight spaces
- On-demand, mid-volume inkjet solution creates photo-quality colour labels in a single pass
- More colour options than you’d ever need—16 million!
- Custom identification without the high cost of outsourced printing
- Includes Brady Workstation Lab Suite to your solution and make label creation even easier
- Setup is stress free – Contact our Brady Technical Support Team and we will schedule a free phone setup appointment
Specifikationer
Specifikationer
| Visningstyp | None |
| Tryckteknik | Inkjet |
| Mått (L x B) | 389 x 264.00 mm |
| Färgkapacitet | Multi-color |
| Inkluderar | This kit contains a J2000 printer, a power cord and AC adapter, a USB cable, a Quick Start Guide, an ink cartridge, a material roll for printhead alignment, a USB stick with drivers and manuals and a Brady Workstation Laboratory Suite |
| Utskriftshastighet | 63.50 mm/sec. |
| Upplösning | 4800 dpi |
| Strömförsörjningsdetaljer | 100/240 V |