Learn More
Brady™ BradyJet™ J2000 Full Color Ink Cartridge
Brady™ BradyJet™ J2000 Full Color Ink Cartridge is a replacement cartridge that makes sure colors stay vibrant.
Brand: Brady™ 150154
1258.93 SEK valid until 2025-06-30
Use promo code "24156" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with the BradyJet™ J2000 Color Label Printer
- Colors: Cyan, Magenta, Yellow
Specifications
Cyan/Magenta/Yellow | |
3.9 x 1.6 x 2.8 in. | |
J20-CMY |
BradyJet J2000 | |
1 Ea. |