Beskrivning
Designed specifically for BRAND™ PCR plates.
- Reduces evaporation losses by up to 75 percent in comparison to conventional systems
- Highly flexible
- Forms tight seal
- Easily pierced with pipet tips.
Specifikationer
Specifikationer
| Material | TPE |
| Autoklaveringsbar | Autoclavable |
| Antal Wells | 96 |
| Antal per förpackning | 5 |
| Produkttyp | Sealing Mat |
| Plattkompatibilitet | 96-well PCR Plate |