Learn More
BRAND™ Pipet Tips, 1 to 50uL
Fits most pipettes that use standard ultra-micro/crystal tips
Brand:  BRAND™ 732006
Additional Details : Weight : 0.46500kg
 View more versions of this product
Product Code. 15184887
Quantity Price
1 231.00 SEK / Pack of 1000
2 230.79 SEK / Pack of 1000
Estimated Shipment
05-03-2020
Add to basket
Description
Description
  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Nonsterile
  • Available in either standard or ultra low retention types
  • Available either in bulk packages or in convenient rack format
  • All unopened packages are DNA-, RNase-, ATP- and endotoxin-free
  • For reverse pipetting of small volumes
Ultra low retention tips
  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface with low surface tension for minimal retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain proteins, nucleic acids, detergents or other difficult liquids where sample loss is possible
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention properties
Specifications
Specifications
Universal Fit
Non-sterile
PP (Polypropylene)
High translucency, easy ejection and low waste, quick volume check
1 to 50μL
Colorless
DNase, RNase, ATP, and Endotoxin Free