Learn More
BrandTech™ BRANDplates™ cellGrade™ premium 384-Well Microplates
For serum-free and serum-reduced cultivation of cells
Brand:  BrandTech™ 782106
Additional Details : Weight : 4.03000kg
 View more versions of this product
Product Code. 10012743
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
View alternative products »
Specifications
Specifications
ANSI, SLAS
Cultivation of fastidious adherent cell lines
Polystyrene
Sterile
Premium 384 -Well Standard Microplate
Flat Bottom
120μL
White
384 well
384
Carboxyl, Hydroxyl and free Amino groups
120μL
With Lid